missing translation for 'onlineSavingsMsg'
Learn More

3-Oxetanone, 95%

Product Code. 15239646 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15239646 5 g 5g
15229646 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15239646 Supplier Thermo Scientific Acros Supplier No. 451200050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6704-31-0 | C3H4O2 | 72.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6704-31-0
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
InChI Key ROADCYAOHVSOLQ-UHFFFAOYSA-N
Synonym 3-oxetanone, 1,3-epoxy-2-propanone, 3-oxooxetane, oxetane-3-one, oxetan-3-on, 1,3-epoxypropanone, pubchem15880, 3-oxentanone, acmc-209nxj, ksc358e8n
PubChem CID 15024254
IUPAC Name oxetan-3-one
SMILES C1C(=O)CO1

Specifications

Boiling Point 140°C
Flash Point 26°C
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 72.06
Percent Purity 95%
Chemical Name or Material 3-Oxetanone
compliance-icons compliance-icons
Product Identifier
  • 3-Oxetanone, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.