missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, Thermo Scientific™

Product Code. 15407116
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15407116 250 mg 250mg
15497106 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15407116

Brand: Thermo Scientific Alfa Aesar H62168.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 943994-02-3
Molecular Formula C14H18BNO4
Molecular Weight (g/mol) 275.111
MDL Number MFCD12755796
InChI Key ZXOSNHPLTJAXSA-UHFFFAOYSA-N
Synonym 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 3-oxo-2h,4h-benzo b 1,4 oxazine-6-boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,4-dihydro-1,4-benzoxazin-3-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 3-oxo-3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2h-benzo b 1,4 oxazin-3 4h-one, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazin-3-ol, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-4h-1,4-benzoxazin-3-one
PubChem CID 46856474
IUPAC Name 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4H-1,4-benzoxazin-3-one
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)OCC(=O)N3

Specifications

Quantity 250 mg
Formula Weight 275.11
Physical Form Solid
Chemical Name or Material 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.