missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11300647
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11300647 250 mg 250mg
10685253 1 g 1g
10792197 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11300647 Supplier Thermo Scientific Maybridge Supplier No. CC46803CB
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 31794-45-3
Molecular Formula C8H6ClNO4S
Molecular Weight (g/mol) 247.649
MDL Number MFCD05664887
InChI Key CGTCULUUVYBAPX-UHFFFAOYSA-N
Synonym 3-oxo-3,4-dihydro-2h-1,4-benzoxazine-6-sulfonyl chloride, 2h-1,4-benzoxazine-6-sulfonyl chloride, 3,4-dihydro-3-oxo, 3-oxo-3,4-dihydro-2h-benzo b 1,4 oxazine-6-sulfonyl chloride, 3,4-dihydro-3-oxo-2h-1,4-benzoxazine-6-sulphonyl chloride, 3-oxo-3,4-dihydro-2h-benzo 1,4 oxazine-6-sulfonyl chloride, 3-oxo-2,4-dihydro-1,4-benzoxazine-6-sulfonyl chloride, 6-chlorosulfonyl-2h,4h-benzo e 1,4-oxazin-3-one, 6-chlorosulphonyl-3-oxo-1,4-benzoxazine, 3-oxo-3,4-dihydro-2h-1,4-benzooxazine-6-sulfonyl chloride, 3-oxo-3,4-dihydro-2h-1,4-benzoxazine-6-sulfonyl ch
PubChem CID 5200229
IUPAC Name 3-oxo-4H-1,4-benzoxazine-6-sulfonyl chloride
SMILES C1C(=O)NC2=C(O1)C=CC(=C2)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 247.66
Percent Purity 97%
Chemical Name or Material 3-Oxo-3, 4-dihydro-2H-1, 4-benzoxazine-6-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.