missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carboxylic Acid, TRC

Product Code. 30382058
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382058 500 mg
30379999 50 mg
30373697 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382058 Supplier Toronto Research Chemicals Supplier No. TRCE588758500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134997-87-8
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.16
Synonym 3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-6-carboxylic Acid, 3-Hydroxy-2H-1,4-benzoxazine-6-carboxylic Acid, 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic Acid, 3-Oxo-3,4-dihydro-2H-benzo[1,4]oxazine-6-carboxylic Acid, 3-Oxo-4H-1,4-benzoxazine-6-carboxylic Acid
IUPAC Name 3-oxo-4H-1,4-benzoxazine-6-carboxylic acid
SMILES OC(=O)c1ccc2OCC(=O)Nc2c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H7NO4/c11-8-4-14-7-2-1-5(9(12)13)3-6(7)10-8/h1-3H,4H2,(H,10,11)(H,12,13)
Formula Weight 193.0375
Chemical Name or Material 3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.