missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile, 97%, Thermo Scientific™

Product Code. 10190782
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10190782 10 g Amber Glass Bottle 10g
10099431 1 g Amber Glass Bottle 1g
10062304 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10190782 Supplier Thermo Scientific Maybridge Supplier No. KM04838EA
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 122454-46-0
Molecular Formula C10H6F3NO2
Molecular Weight (g/mol) 229.16
MDL Number MFCD00052146
InChI Key IYZQGBBCANKWMX-UHFFFAOYSA-N
Synonym 3-oxo-3-4-trifluoromethoxy phenyl propanenitrile, 4-trifluoromethoxy benzoyl acetonitrile, 4-trifluoromethoxy benzoylacetonitrile, 4-trifluoromethoxybenzoyl acetonitrile, 3-oxo-3-4-trifluoromethoxy phenyl propionitrile, benzenepropanenitrile, beta-oxo-4-trifluoromethoxy, maybridge1_004897, acmc-1bxb1
PubChem CID 2777239
IUPAC Name 3-oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile
SMILES FC(F)(F)OC1=CC=C(C=C1)C(=O)CC#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CF3OC6H4C(O)CH2CN
Quantity 10 g
Formula Weight 229.16
Percent Purity 97%
Chemical Name or Material 3-Oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.