missing translation for 'onlineSavingsMsg'
Learn More

3-Oxobutanenitrile, 96%

Product Code. 15481799
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15481799 250 mg 250mg
15471799 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15481799 Supplier Thermo Scientific Alfa Aesar Supplier No. H64536.MD
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2469-99-0 | C4H5NO | 83.09 g/mol

It is used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2469-99-0
Molecular Formula C4H5NO
Molecular Weight (g/mol) 83.09
MDL Number MFCD05861381
InChI Key OPXYNEYEDHAXOM-UHFFFAOYSA-N
Synonym 3-oxobutyronitrile, 3-oxo-butyronitrile, butanenitrile, 3-oxo, cyanoacetone, cyanaceton, a-cyanoacetone, acetoacetonitrile, acetylacetonitrile, 1-cyano-2-propanone, alpha-acetylacetonitrile
PubChem CID 75579
IUPAC Name 3-oxobutanenitrile
SMILES CC(=O)CC#N

Specifications

Color Colorless to Yellow
Boiling Point 92°C to 94°C (10 mmHg)
Linear Formula CH3COCH2CN
Refractive Index 1.4285
Quantity 250 mg
UN Number UN3276
Beilstein 1633584
Solubility Information Slightly soluble in water.
Formula Weight 83.09
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Oxobutanenitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.