missing translation for 'onlineSavingsMsg'
Learn More

3-Oxocyclobutanecarboxylic acid, 98%

Product Code. 15472119
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15472119 5 g 5g
15482119 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15472119 Supplier Thermo Scientific Alfa Aesar Supplier No. H64774.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 23761-23-1 | C5H6O3 | 114.10 g/mol

Useful intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23761-23-1
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.10
MDL Number MFCD00100900
InChI Key IENOFRJPUPTEMI-UHFFFAOYSA-N
Synonym 3-oxocyclobutanecarboxylic acid, 3-oxo-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-oxo, 3-oxo-cyclobutanecarboxylicacid, 1-carboxy-3-oxocyclobutane, 3-oxocyclobutyl carboxylic acid, 3-oxocyclobutanecarboxylicacid, pubchem14220, acmc-1cak7, ksc201q6h
PubChem CID 4913358
IUPAC Name 3-oxocyclobutane-1-carboxylic acid
SMILES OC(=O)C1CC(=O)C1

Specifications

Melting Point 69°C to 70°C
Color White
Quantity 5 g
Beilstein 2078401
Solubility Information Slightly soluble in water.
Formula Weight 114.1
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Oxocyclobutanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.