missing translation for 'onlineSavingsMsg'
Learn More

3-Oxotetrahydrofuran, TRC

Product Code. 30425853
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425853 5 g 5g
30423489 1 g -
30417470 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425853 Supplier Toronto Research Chemicals Supplier No. TRCO8704805G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 22929-52-8
Molecular Formula C4 H6 O2
Molecular Weight (g/mol) 86.09
Synonym 4,5-Dihydro-3(2H)-furanone, Dihydro-3(2H)-furanone, Dihydrofuran-3-one, Oxolan-3-one
IUPAC Name oxolan-3-one
SMILES O=C1CCOC1

Especificaciones

Quantity 5 g
InChI Formula InChI=1S/C4H6O2/c5-4-1-2-6-3-4/h1-3H2
Formula Weight 86.0368
Chemical Name or Material 3-Oxotetrahydrofuran
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.