Learn More
TERUMO 3-Part Syringe
Product Code. 31924915
Change view
Click to view available options
Volume (Metric):
1 mL
Unit Size:
Pack of 100
Product Code. Volume (Metric) Unit Size
31924915 1 mL Pack of 100
1 options
Product Code. 31924915 Supplier TERUMO Supplier No. 8MDSS01LE
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Needleless syringe with siliconization for optimal plunger mobility.
  • The integral gasket ensures a firm and distinct end-stop
  • Clear graduated scale allows accurate dosage
  • Transparent barrel for visualisation
  • Finger grips to support easy handling
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance Class IIa Medical Device
Material Polypropylene (Barrel), Polypropylene (Plunger)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Syringe
Disposable Single-use
Volume (Metric) 1 mL
Connection Type Centric Luer Lock
Graduated Yes
Sterility Sterile