missing translation for 'onlineSavingsMsg'
Learn More

3-Pentanone, ReagentPlus™, ≥99.0% (GC), Honeywell™

Product Code. 15621190 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
Unit Size:
1 L
100mL
2.5 L
20 L
500mL
Product Code. Quantity unitSize
15621190 100 mL 100mL
15671190 500 mL 500mL
15641190 1 L 1 L
15651190 2.5 L 2.5 L
15661190 20 L 20 L
5 options
This item is not returnable. View return policy

Product Code. 15621190

Brand: Honeywell Research Chemicals 127604100ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99.0% (GC)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-22-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00009320
InChI Key FDPIMTJIUBPUKL-UHFFFAOYSA-N
Synonym 3-pentanone, diethyl ketone, propione, ethyl ketone, dimethylacetone, metacetone, methacetone, diethylcetone, ethyl propionyl, diethylketone
PubChem CID 7288
ChEBI CHEBI:67886
IUPAC Name pentan-3-one
SMILES CCC(=O)CC

Specifications

Melting Point -39°C
Color Colorless
Density 0.813g/mL at 25°C
Boiling Point 102°C
Flash Point 7°C (44.6°F)
Packaging Glass Bottle
Linear Formula CH3CH2COCH2CH3
Refractive Index n20/D 1.392(lit.)
Quantity 100 mL
UN Number UN1156
Beilstein 635749
Solubility Information Slightly soluble in water
Formula Weight 86.13g/mol
Percent Purity ≥99.0% (GC)
Grade ReagentPlus
Chemical Name or Material 3-Pentanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.