missing translation for 'onlineSavingsMsg'
Learn More

3-Pentanone, TRC

Product Code. 30409457
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30409457 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30409457

Brand: Toronto Research Chemicals TRCP27360025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Pentanone
CAS 96-22-0
Molecular Formula C5 H10 O
Quantity 25 g
Synonym 3-Pentanone, DEK, Diethyl ketone, Dimethylacetone, Ethyl ketone, Metacetone, Methacetone, NSC 8653, Propione, ZED 2EK, Pentan-3-one
InChI Formula InChI=1S/C5H10O/c1-3-5(6)4-2/h3-4H2,1-2H3
SMILES CCC(=O)CC
IUPAC Name pentan-3-one
Molecular Weight (g/mol) 86.13
Formula Weight 86.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.