missing translation for 'onlineSavingsMsg'
Learn More

3-Penten-2-one, 70%, technical

Product Code. 10861851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10861851 1 g Glass bottle 1g
10179629 5 g Glass bottle 5g
10068269 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10861851 Supplier Thermo Scientific Acros Supplier No. 163290010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 625-33-2 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-33-2
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00009290
InChI Key LABTWGUMFABVFG-ONEGZZNKSA-N
Synonym z-pent-3-en-2-one, unii-w8xx21whyc, z-3-penten-2-one, w8xx21whyc, 3-penten-2-one, 3z-pent-3-en-2-one, 3-penten-2-one, z, cis-3-penten-2-one, 3-penten-2-one, cis
PubChem CID 637920
IUPAC Name (E)-pent-3-en-2-one
SMILES CC=CC(=O)C

Specifications

CAS Min % 65.0
CAS Max % 100.0
Color Colorless
Density 0.8600g/mL
Boiling Point 121.0°C to 124.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 70% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHCOCH3
Refractive Index 1.4405
Quantity 1 g
Beilstein 01,732
Fieser 02,306; 03,218; 04,371
Specific Gravity 0.86
Formula Weight 84.12
Percent Purity 70%
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-Penten-2-one
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Penten-2-one, 70%, Technical Grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.