missing translation for 'onlineSavingsMsg'
Learn More

3-Penten-2-one, tech. 85%

Product Code. 11307856
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307856 5 g 5g
11397846 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307856 Supplier Thermo Scientific Alfa Aesar Supplier No. L13031.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-33-2 | C5H8O | 84.118 g/mol

3-Penten-2-one

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Penten-2-one

Solubility
Slightly soluble in Water

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with bases, reducing agents and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-33-2
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00009290
InChI Key LABTWGUMFABVFG-ONEGZZNKSA-N
Synonym z-pent-3-en-2-one, unii-w8xx21whyc, z-3-penten-2-one, w8xx21whyc, 3-penten-2-one, 3z-pent-3-en-2-one, 3-penten-2-one, z, cis-3-penten-2-one, 3-penten-2-one, cis
PubChem CID 637920
IUPAC Name (E)-pent-3-en-2-one
SMILES CC=CC(=O)C

Specifications

Density 0.862
Boiling Point 119°C to 121°C
Flash Point 21°C (69°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.4355
Quantity 5 g
UN Number UN1992
Beilstein 1633505
Solubility Information Slightly soluble in Water
Formula Weight 84.12
Percent Purity 85%
Grade Technical
Chemical Name or Material 3-Penten-2-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.