missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenoxymethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10162934
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10162934 250 mg Amber Glass Bottle 250mg
10328883 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10162934

Brand: Thermo Scientific Maybridge CC63604CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 104508-27-2
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD09025913
InChI Key KZUJGQAFTCDBKA-UHFFFAOYSA-N
Synonym 3-phenoxymethyl benzaldehyde, benzaldehyde,3-phenoxymethyl, acmc-1cada
PubChem CID 21288517
IUPAC Name 3-(phenoxymethyl)benzaldehyde
SMILES C1=CC=C(C=C1)OCC2=CC=CC(=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.25
Percent Purity 97%
Chemical Name or Material 3-(Phenoxymethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.