missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenoxymethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10328883
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10328883 1 g Amber Glass Bottle 1g
10162934 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10328883 Supplier Thermo Scientific Maybridge Supplier No. CC63604DA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 104508-27-2
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD09025913
InChI Key KZUJGQAFTCDBKA-UHFFFAOYSA-N
Synonym 3-phenoxymethyl benzaldehyde, benzaldehyde,3-phenoxymethyl, acmc-1cada
PubChem CID 21288517
IUPAC Name 3-(phenoxymethyl)benzaldehyde
SMILES C1=CC=C(C=C1)OCC2=CC=CC(=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 212.25
Percent Purity 97%
Chemical Name or Material 3-(Phenoxymethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.