missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxypropyl Bromide, TRC

Product Code. 30414941
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414941 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414941 Supplier Toronto Research Chemicals Supplier No. TRCP31887025G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 588-63-6
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
Synonym 3-Bromopropoxy)benzene, 1-Bromo-3-phenoxypropane, 3-Bromo-1-phenoxypropane3-Bromopropyl Phenyl Ether, 3-Phenoxy-1-bromopropane, 3-Phenoxypropyl BromideNSC 2641, gamma-Phenoxypropyl Bromide
IUPAC Name 3-bromopropoxybenzene
SMILES BrCCCOc1ccccc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H11BrO/c10-7-4-8-11-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2
Formula Weight 213.9993
Chemical Name or Material 3-Phenoxypropyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.