missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxypropylamine hydrochloride, 97+%, Thermo Scientific™

Product Code. 15459107
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459107 250 mg 250mg
15449107 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459107 Supplier Thermo Scientific Alfa Aesar Supplier No. H52398.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83708-39-8
Molecular Formula C9H14ClNO
Molecular Weight (g/mol) 187.67
MDL Number MFCD01325325
InChI Key JTWGNQQRTPIYSJ-UHFFFAOYSA-N
Synonym 3-aminopropoxy benzene hydrochloride, 3-phenoxypropan-1-amine hydrochloride, 3-phenoxypropylamine hydrochloride, 3-aminopropoxy benzene hcl, 3-phenoxypropylamine, chloride, 3-phenoxypropan-1-aminehydrochloride, 3-phenoxypropyl amine hydrochloride, 3-phenoxypropan-1-amine hcl, 3-aminopropoxy benzenehydrochloride
PubChem CID 18590545
IUPAC Name 3-phenoxypropan-1-amine hydrochloride
SMILES Cl.NCCCOC1=CC=CC=C1

Specifications

Linear Formula C6H5O(CH2)3NH2·HCl
Quantity 250 mg
Beilstein 3697706
Formula Weight 187.67
Percent Purity ≥97%
Chemical Name or Material 3-Phenoxypropylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.