missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1H-benzo[f]chromen-1-one, 98%, BLD Pharm™

Product Code. 31107084
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31107084 250 mg
31096268 1 g
31106166 5 g
31098089 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31107084 Supplier BLD Pharm Supplier No. BD00827527250mg
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6051-87-2
Molecular Formula C19H12O2
MDL Number MFCD00004986
Quantity 250 mg
SMILES O=C1C=C(OC2=CC=C3C(C=CC=C3)=C21)C4=CC=CC=C4
Molecular Weight (g/mol) 272.30
Percent Purity 98%
Chemical Name or Material 3-Phenyl-1H-benzo[f]chromen-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.