missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™

Product Code. 10039271
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10039271 1 g Amber Glass Bottle 1g
10049271 10 g Amber Glass Bottle 10g
10196782 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10039271 Supplier Thermo Scientific Maybridge Supplier No. KM00060DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1572-10-7
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
InChI Key PWSZRRFDVPMZGM-UHFFFAOYSA-N
Synonym 3-phenyl-1h-pyrazol-5-amine, 3-amino-5-phenylpyrazole, 5-amino-3-phenylpyrazole, 1h-pyrazol-3-amine, 5-phenyl, 5-phenyl-2h-pyrazol-3-amine, pyrazole, 5-amino-3-phenyl, 3-amino-5-phenyl-1h-pyrazole, 5-phenyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 3-phenyl, 5-phenyl-1h-pyrazol-3-ylamine
PubChem CID 136655
IUPAC Name 5-phenyl-1H-pyrazol-3-amine
SMILES C1=CC=C(C=C1)C2=CC(=NN2)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 5-Phenyl-1H-pyrazol-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.