missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10177142
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10177142 1 g Amber Glass Bottle 1g
10115832 250 mg Amber Glass Bottle 250mg
10184244 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10177142 Supplier Thermo Scientific Maybridge Supplier No. MO01317DA
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1134-49-2
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD01248821,MFCD05170017
InChI Key QBPUOAJBMXXBNU-UHFFFAOYSA-N
Synonym 5-phenyl-1h-pyrazole-3-carboxylic acid, 5-phenyl-2h-pyrazole-3-carboxylic acid, 3-phenylpyrazole-5-carboxylic acid, pyrazole-5-carboxylic acid, 3-phenyl, 1h-pyrazole-3-carboxylic acid, 5-phenyl, 5-phenyl-1h-pyrazole-3-carboxylicacid, pyrazole-3-carboxylic acid, 5-phenyl, 1h-pyrazole-3-carboxylicacid, 5-phenyl, 1h-pyrazole-5-carboxylicacid, 3-phenyl, tos-bb-0266
PubChem CID 121025
IUPAC Name 3-phenyl-1H-pyrazole-5-carboxylic acid
SMILES OC(=O)C1=CC(=NN1)C1=CC=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 5-Phenyl-1H-pyrazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.