missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-2-propyn-1-ol, 98+%

Product Code. 11360418
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11360418 2 g 2g
11350418 10 g 10g
11340418 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11360418 Supplier Thermo Scientific Alfa Aesar Supplier No. 043778.04
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1504-58-1 | C9H8O | 132.162 g/mol

3-Phenyl-2-propyn-1-ol is an phenyl acetylene derivative with inhibitory effect on nitrification in soil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1504-58-1
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00040914
InChI Key NITUNGCLDSFVDL-UHFFFAOYSA-N
Synonym 3-phenyl-2-propyn-1-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-prop-2-yn-1-ol, 1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, pubchem13580, 3-phenyl-2-propyn1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, 1-phenyl-propyn-3-ol
PubChem CID 123115
IUPAC Name 3-phenylprop-2-yn-1-ol
SMILES C1=CC=C(C=C1)C#CCO

Specifications

Density 1.078 g/mL
Boiling Point 129°C to 130°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5867
Quantity 2 g
Beilstein 2040911
Solubility Information Soluble in ether,acetone,benzene,chloroform,ethyl Acetate,methanol.
Formula Weight 132.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 3-Phenyl-2-propyn-1-ol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.