missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylisoxazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10706725
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10706725 5 g Amber Glass Bottle 5g
10396704 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10706725 Supplier Thermo Scientific Maybridge Supplier No. CC64501DE
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 14442-12-7
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD01936010
InChI Key YGTFDJRCCBKLDM-UHFFFAOYSA-N
Synonym 3-phenylisoxazole-5-carboxylic acid, 3-phenyl-5-isoxazolecarboxylic acid, 5-isoxazolecarboxylic acid, 3-phenyl, 3-phenyl-isoxazole-5-carboxylic acid, phenylisoxazolecarboxylicacid, 5-carboxy-3-phenylisoxazole, 3-phenylisoxazole-5-carboxylic, 3-phenyl-5-isoxazolecarboxylicacid, 3-phenylisoxazole-5-carboxylicacid, 3-phenyl-5-isoxazole carboxylic acid
PubChem CID 737474
IUPAC Name 3-phenyl-1,2-oxazole-5-carboxylic acid
SMILES C1=CC=C(C=C1)C2=NOC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 3-Phenylisoxazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.