missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionyl Chloride, TRC

Product Code. 30424840
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424840 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424840 Supplier Toronto Research Chemicals Supplier No. TRCP33621525G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Phenylpropionyl Chloride
CAS 645-45-4
Molecular Formula C9H9ClO
Quantity 25 g
Synonym Benzenepropanoyl Chloride, Hydrocinnamoyl Chloride, 3-Phenylpropanoic Acid Chloride, 3-Phenylpropanoyl Chloride, 3-Phenylpropionic Acid Chloride, Dihydrocinnamoyl Chloride, Hydrocinnamyl Chloride, NSC 2854, β-Phenylpropanoyl Chloride, β-Phenylpropionyl Chloride
InChI Formula InChI=1S/C9H9ClO/c10-9(11)7-6-8-4-2-1-3-5-8/h1-5H,6-7H2
SMILES ClC(=O)CCc1ccccc1
IUPAC Name 3-phenylpropanoyl chloride
Molecular Weight (g/mol) 168.62
Formula Weight 168.0342
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.