missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropyl isocyanate, 97%, Thermo Scientific™

Product Code. 15545966 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15545966 1 g 1g
15555966 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15545966 Supplier Thermo Scientific Acros Supplier No. 457380010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 68664-23-3
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.21
InChI Key WPABAKMRZCBXTO-UHFFFAOYSA-N
Synonym 3-phenylpropyl isocyanate, 3-phenylpropylisocyanate, 3-isocyanatopropyl benzene, phenylpropylisocyanate, acmc-20amyq, 3-isocyanatopropyl-benzene, 1-3-isocyanatopropyl benzene, 3-isocyanato-propyl-benzene, benzene, 3-isocyanatopropyl
PubChem CID 3427666
IUPAC Name 3-isocyanatopropylbenzene
SMILES C1=CC=C(C=C1)CCCN=C=O

Specifications

Boiling Point 115.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 161.21
Percent Purity 97%
Chemical Name or Material 3-Phenylpropyl isocyanate
compliance-icons compliance-icons
Product Identifier
  • 3-Phenylpropyl isocyanate, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.