missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15405385
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405385 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15405385 Supplier Thermo Scientific Alfa Aesar Supplier No. H58569.AD
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 626207-42-9 | C9H11BrZn | 264.471 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Tekniske data

CAS 626207-42-9
Molecular Formula C9H11BrZn
Linear Formula C6H5CH2CH2CH2ZnBr
MDL Number MFCD08064069
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym 3-phenyl-1-propylzinc bromide, bromo 3-phenylpropyl zinc, 3-phenyl-1-propylzinc bromide 0.5 m in tetrahydrofuran, 3-phenylpropylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key XTDLRSRUPYQIGX-UHFFFAOYSA-M
SMILES [CH2-]CCC1=CC=CC=C1.[Zn+]Br
IUPAC Name bromozinc(1+);propylbenzene
Molecular Weight (g/mol) 264.471
PubChem CID 24722663
Formula Weight 264.47
Chemical Name or Material 3-Phenylpropylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.