missing translation for 'onlineSavingsMsg'
Learn More

3-Picoline, 99%

Product Code. 10051480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051480 250 mL Glass bottle 250mL
10347110 500 mL Glass bottle 500mL
10387060 1 L Glass bottle 1 L
10246813 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10051480 Supplier Thermo Scientific Acros Supplier No. 131132500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-99-6 | C6H7N | 93.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-99-6
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00006402
InChI Key ITQTTZVARXURQS-UHFFFAOYSA-N
Synonym 3-picoline, beta-picoline, pyridine, 3-methyl, m-picoline, m-methylpyridine, b-picoline, beta-methylpyridine, .beta.-methylpyridine, .beta.-picoline, meta-methylpyridine
PubChem CID 7970
ChEBI CHEBI:39922
IUPAC Name 3-methylpyridine
SMILES CC1=CN=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -19.0°C
Color Yellow
Density 0.9500g/mL
Boiling Point 143.0°C to 144.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5050 to 1.5070
Quantity 250 mL
Beilstein 20, 239
Merck Index 15, 7513
Specific Gravity 0.95
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 93.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Picoline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Picoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 3-Methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.