missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Picoline N-oxide, 98%

Product Code. 11444284
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11444284

Brand: Alfa Aesar™ A12578.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1003-73-2
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00006203
InChI Key DMGGLIWGZFZLIY-UHFFFAOYSA-N
Synonym 3-methylpyridine 1-oxide, 3-picoline-n-oxide, 3-picoline n-oxide, 3-methylpyridine n-oxide, 3-methylpyridine-n-oxide, pyridine, 3-methyl-, 1-oxide, 3-picoline 1-oxide, beta-picoline n-oxide, 3-methylpyridin-1-ium-1-olate, 3-methylpyridine-1-oxide
PubChem CID 13858
IUPAC Name 3-methyl-1-oxidopyridin-1-ium
SMILES CC1=C[N+](=CC=C1)[O-]

Specifications

Melting Point 36°C to 40°C
Density 1.13
Boiling Point 150°C (15 mmHg)
Flash Point 155°C (311°F)
Quantity 100 g
Beilstein 106327
Sensitivity Hygroscopic
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 3-Picoline N-oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.