missing translation for 'onlineSavingsMsg'
Learn More

3-(Piperidin-1-ylmethyl)benzoic acid hydrochloride hemihydrate, ≥97%, Thermo Scientific™

Product Code. 10669934
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10669934 250 mg Amber Glass Bottle 250mg
10351143 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10669934 Supplier Thermo Scientific Maybridge Supplier No. CC45251CB
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 863991-96-2
Molecular Formula C26H38Cl2N2O5
Molecular Weight (g/mol) 529.50
MDL Number MFCD07772860
InChI Key HTWIFQGKFFGIFX-UHFFFAOYSA-N
Synonym 3-piperdin-1-ylmethyl benzoic acid hydrochloride hemihydrate, bis 3-piperidin-1-ylmethyl benzoic acid hydrate dihydrochloride, benzoic acid,3-1-piperidinylmethyl-,hydrochloride,hydrate 2:2:1, 3-piperidin-1-yl methyl benzoic acid-hydrogen chloride-water 2/2/1
PubChem CID 44119333
IUPAC Name 3-(piperidin-1-ylmethyl)benzoic acid;hydrate;dihydrochloride
SMILES O.Cl.Cl.OC(=O)C1=CC(CN2CCCCC2)=CC=C1.OC(=O)C1=CC(CN2CCCCC2)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 264.76
Percent Purity 97%
Chemical Name or Material 3-(Piperidinomethyl)benzoic acid hydrochloride 0.5 hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.