missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinobenzoic acid, 97%, Thermo Scientific™

Product Code. 10620725
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10620725 1 g Amber Glass Bottle 1g
10461323 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10620725 Supplier Thermo Scientific Maybridge Supplier No. CC34001DA
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 77940-94-4
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
MDL Number MFCD06659081
InChI Key FLQRORYAJSTYLT-UHFFFAOYSA-N
Synonym 3-piperidinobenzoic acid, 3-piperidin-1-yl benzoic acid, 3-piperidin-1-yl-benzoic acid, 1-3-carboxyphenyl piperidine, 3-1-piperidinyl benzoic acid, 3-piperidylbenzoic acid, benzoic acid,3-1-piperidinyl, benzoic acid, 3-1-piperidinyl
PubChem CID 2795552
IUPAC Name 3-piperidin-1-ylbenzoic acid
SMILES C1CCN(CC1)C2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.26
Percent Purity 97%
Chemical Name or Material 3-Piperidinobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.