missing translation for 'onlineSavingsMsg'
Learn More

3-(Piperidinomethyl)benzaldehyde, ≥97%, Thermo Scientific™

Product Code. 10736715
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10736715 5 g 5g
10027852 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10736715 Supplier Thermo Scientific Maybridge Supplier No. CC45204DE
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 471930-01-5
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
MDL Number MFCD07782418
InChI Key UJDTVJKIOWAUNQ-UHFFFAOYSA-N
Synonym 3-piperidinomethyl benzaldehyde, 3-piperidin-1-ylmethyl benzaldehyde, 3-piperidylmethyl benzaldehyde, 3-1-piperidinylmethyl benzaldehyde, 3-1-piperidinyl methylbenzaldehyde, 1-3-carboxaldehydebenzyl piperidine
PubChem CID 7164643
IUPAC Name 3-(piperidin-1-ylmethyl)benzaldehyde
SMILES C1CCN(CC1)CC2=CC=CC(=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 203.28
Percent Purity 97%
Chemical Name or Material 3-(Piperidinomethyl)benzaldehyde
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.