missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinopyridine-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10163454
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163454 250 mg Amber Glass Bottle 250mg
10031004 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163454 Supplier Thermo Scientific Maybridge Supplier No. CC37601CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-01-5
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD09025831
InChI Key QIWMAMIMWHCYGZ-UHFFFAOYSA-N
Synonym 3-piperidinopyridine-2-carboxylic acid, 3-piperidin-1-yl pyridine-2-carboxylic acid, 3-piperidin-1-yl picolinic acid, 3-piperidylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 3-1-piperidinyl
PubChem CID 18525731
IUPAC Name 3-piperidin-1-ylpyridine-2-carboxylic acid
SMILES C1CCN(CC1)C2=C(N=CC=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 3-Piperidinopyridine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.