missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridineacetic acid hydrochloride, 98+%

Product Code. 11467457
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467457 5 g 5g
11477457 25 g 25g
11487457 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467457 Supplier Thermo Scientific Alfa Aesar Supplier No. A15599.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6419-36-9 | C7H8ClNO2 | 173.596 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6419-36-9
Molecular Formula C7H8ClNO2
Molecular Weight (g/mol) 173.596
MDL Number MFCD00012819
InChI Key XVCCOEWNFXXUEV-UHFFFAOYSA-N
Synonym 3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride
PubChem CID 2723724
IUPAC Name 2-pyridin-3-ylacetic acid;hydrochloride
SMILES C1=CC(=CN=C1)CC(=O)O.Cl

Specifications

Melting Point 158°C to 162°C
Quantity 5 g
Beilstein 3696630
Merck Index 14,7971
Formula Weight 173.6
Percent Purity ≥98%
Chemical Name or Material 3-Pyridineacetic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.