missing translation for 'onlineSavingsMsg'
Learn More

3-Pyridylacetic acid hydrochloride, 98%

Product Code. 10066253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
25 g
Packaging:
Glass bottle
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10066253 25 g Glass bottle 25g
10061480 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10066253 Supplier Thermo Scientific Acros Supplier No. 131960250
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6419-36-9 | C7H7NO2·ClH | 173.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6419-36-9
Molecular Formula C7H7NO2·ClH
Molecular Weight (g/mol) 173.6
MDL Number MFCD00012819
InChI Key XVCCOEWNFXXUEV-UHFFFAOYSA-N
Synonym 3-pyridylacetic acid hydrochloride, 2-pyridin-3-yl acetic acid hydrochloride, pyridine-3-acetic acid hydrochloride, 3-pyridineacetic acid hydrochloride, 3-pyridylacetic acid hcl, pyridin-3-ylacetic acid hydrochloride, 3-pyridineacetic acid hcl, 3-pyridinylacetic acid hcl, pyridine-3-acetic acid hcl, 2-3-pyridinyl acetic acid hydrochloride
PubChem CID 2723724
IUPAC Name 2-pyridin-3-ylacetic acid;hydrochloride
SMILES C1=CC(=CN=C1)CC(=O)O.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 160°C to 161°C
Color Beige or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C5H4N)CH2OOH·HCl
Quantity 25 g
Merck Index 10, 4635
Solubility Information Solubility in water: soluble
Formula Weight 173.6
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 3-Pyridylacetic acid hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • pyridin-3-ylacetic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.