missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-5-ylbenzaldehyde, 97%, Thermo Scientific™

Product Code. 10600335
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10600335 1 g 1g
10525953 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10600335 Supplier Thermo Scientific Maybridge Supplier No. CC41704DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 640769-70-6
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.198
MDL Number MFCD05864833
InChI Key RAHJVJBPRWJPHE-UHFFFAOYSA-N
Synonym 3-pyrimidin-5-yl benzaldehyde, 3-pyrimidin-5-yl-benzaldehyde, 3-5-pyrimidinyl benzaldehyde, benzaldehyde, 3-5-pyrimidinyl, acmc-1b2vh, benzaldehyde,3-5-pyrimidinyl
PubChem CID 4187249
IUPAC Name 3-pyrimidin-5-ylbenzaldehyde
SMILES C1=CC(=CC(=C1)C=O)C2=CN=CN=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 184.2
Percent Purity 97%
Chemical Name or Material 3-Pyrimidin-5-ylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.