missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrrolidin-1-ylaniline, 97%, Thermo Scientific™

Product Code. 10586714
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10586714 10 g Amber Glass Bottle 10g
10177812 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10586714 Supplier Thermo Scientific Maybridge Supplier No. CC38514EA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 115833-93-7
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
InChI Key DNPBFTQZONVQDX-UHFFFAOYSA-N
PubChem CID 1518808
IUPAC Name 3-pyrrolidin-1-ylaniline
SMILES C1CCN(C1)C2=CC(=CC=C2)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 3-pyrrolidin-1-yl aniline, 3-pyrrolidin-1-yl-phenylamine, 3-1-pyrrolidinyl aniline, benzenamine,3-1-pyrrolidinyl, benzenamine, 3-1-pyrrolidinyl, 3-pyrrolidinylphenylamine, 3-pyrrolizinoaniline, acmc-20e1pl, 1-3-aminophenyl pyrrolidine
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 3-Pyrrolidin-1-ylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.