missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrrolidin-1-ylbenzaldehyde, ≥95%, Thermo Scientific™

Product Code. 10351563
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10351563 250 mg 250mg
10779953 1 g 1g
10225714 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10351563 Supplier Thermo Scientific Maybridge Supplier No. CC38504CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-89-7
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD07772825
InChI Key KELYFFMWYRNGTF-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-yl benzaldehyde, 3-1-pyrrolidinyl benzaldehyde, benzaldehyde,3-1-pyrrolidinyl, 3-pyrrolidinylbenzaldehyde
PubChem CID 7164600
SMILES O=CC1=CC(=CC=C1)N1CCCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name 3-pyrrolidin-1-ylbenzaldehyde
Formula Weight 175.23
Percent Purity 95%
Chemical Name or Material 3-Pyrrolidin-1-ylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.