missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride, 90%, Thermo Scientific™

Product Code. 10679515
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10679515 5 g 5g
10071974 250 mg 250mg
10255664 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10679515 Supplier Thermo Scientific Maybridge Supplier No. CC55751DE
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 887922-93-2
Molecular Formula C12H16ClNO2
Molecular Weight (g/mol) 241.715
InChI Key BORITTZEFSYNFT-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-ylmethyl benzoic acid hydrochloride, 3-1-pyrrolidylmethyl benzoic acid hydrochloride, 3-pyrrolidinylmethyl benzoic acid, chloride, 3-1-pyrrolidylmethyl benzoicacidhydrochloride, 3-pyrrolidin-1-ylmethyl-benzoic acid hydochloride, 3-pyrrolidin-1-ylmethyl-benzoic acid hydrochloride, 3-pyrrolidin-1-yl methyl benzoic acid-hydrogen chloride 1/1
PubChem CID 18525874
IUPAC Name 3-(pyrrolidin-1-ylmethyl)benzoic acid;hydrochloride
SMILES C1CCN(C1)CC2=CC=CC(=C2)C(=O)O.Cl

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 241.72
Percent Purity 90%
Chemical Name or Material 3-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.