missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrroline, 96%

Product Code. 11399115
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399115 1 g 1g
11319125 250 mg 250mg
11309125 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399115 Supplier Thermo Scientific Alfa Aesar Supplier No. L16349.03
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-96-6 | C4H7N | 69.107 g/mol

3-Pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It may be used in the synthesis of renin inhibitors2 and vasodilators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It may be used in the synthesis of renin inhibitors2 and vasodilators.

Solubility
Fully misicble in water.

Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-96-6
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00005213
InChI Key JVQIKJMSUIMUDI-UHFFFAOYSA-N
PubChem CID 66059
ChEBI CHEBI:20198
IUPAC Name 2,5-dihydro-1H-pyrrole
SMILES C1C=CCN1

Specifications

Density 0.902
Boiling Point 90°C to 91°C
Flash Point −18°C (0°F)
Refractive Index 1.469
Quantity 1 g
UN Number UN2733
Beilstein 103173
Sensitivity Hygroscopic
Merck Index 14,8017
Synonym 3-pyrroline, 2,5-dihydropyrrole, 1h-pyrrole, 2,5-dihydro, unii-n0kyf81sdy, delta 3-pyrroline, .delta.3-pyrroline, n0kyf81sdy, delta3-pyrroline, 2,5-dihydropyrrol, 2,5-dihydro pyrrole
Solubility Information Fully misicble in water.
Formula Weight 69.11
Percent Purity 96%
Chemical Name or Material 3-Pyrroline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.