missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrroline, tech. 85% (remainder pyrrolidine)

Product Code. 11302019
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302019 1 g 1g
11312019 5 g 5g
11322019 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11302019 Supplier Thermo Scientific Alfa Aesar Supplier No. H26072.03
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 109-96-6 | C4H7N | 69.107 g/mol

3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Pyrroline is suitable to the core-level binding energies of simple unsaturated organic molecules bonded to the Si(001) surface by X-ray photoelectron spectroscopy (XPS). It is used in the synthesis of renin inhibitors and vasodilators.

Solubility
Soluble in alcohol and ether. Miscible with water.

Notes
Air sensitive. Store at -20°C. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-96-6
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00005213
InChI Key JVQIKJMSUIMUDI-UHFFFAOYSA-N
PubChem CID 66059
ChEBI CHEBI:20198
IUPAC Name 2,5-dihydro-1H-pyrrole
SMILES C1C=CCN1

Specifications

Density 0.902
Boiling Point 90°C to 91°C
Flash Point −18°C (0°F)
Odor Unpleasant
Refractive Index 1.469
Quantity 1 g
UN Number UN2733
Beilstein 103173
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,8017
Synonym 3-pyrroline, 2,5-dihydropyrrole, 1h-pyrrole, 2,5-dihydro, unii-n0kyf81sdy, delta 3-pyrroline, .delta.3-pyrroline, n0kyf81sdy, delta3-pyrroline, 2,5-dihydropyrrol, 2,5-dihydro pyrrole
Solubility Information Soluble in alcohol and ether. Miscible with water.
Formula Weight 69.11
Percent Purity 85%
Grade Technical
Assay (remainder pyrrolidine)
Chemical Name or Material 3-Pyrroline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.