missing translation for 'onlineSavingsMsg'
Learn More

3-Quinolineboronic acid pinacol ester, 97%

Product Code. 11976501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11976501 1 g 1g
11986501 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976501 Supplier Thermo Scientific Acros Supplier No. 432110010
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 171364-85-5 | C15H18BNO2 | 255.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171364-85-5
Molecular Formula C15H18BNO2
Molecular Weight (g/mol) 255.12
MDL Number MFCD05155218
InChI Key ARRJAONCYUAPJR-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid pinacol ester, 3-quinolineboronic acid pinacol ester, quinoline-3-boronic acid pinacolate, 3-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid, pinacol ester, quinoline, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem1882, quinoline-3-boronic acidpinacolate, quinoline-3-boronicacidpinacolester
PubChem CID 10848590
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
SMILES CC1(C)OB(OC1(C)C)C1=CC2=CC=CC=C2N=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 55°C to 57°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 255.13
Percent Purity 97%
Physical Form Powder or Solid
Chemical Name or Material 3-Quinolineboronic acid pinacol ester
Show More Show Less
compliance-icons
Product Identifier
  • 3-Quinolineboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.