missing translation for 'onlineSavingsMsg'
Learn More

3-Quinolinecarboxylic Acid, TRC

Product Code. 30410084
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410084 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410084 Supplier Toronto Research Chemicals Supplier No. TRCQ7005001G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Quinolinecarboxylic Acid
CAS 6480-68-8
Molecular Formula C10 H7 N O2
Quantity 1 g
Synonym 3-Carboxyquinoline, NSC 403263
InChI Formula InChI=1S/C10H7NO2/c12-10(13)8-5-7-3-1-2-4-9(7)11-6-8/h1-6H,(H,12,13)
SMILES OC(=O)c1cnc2ccccc2c1
IUPAC Name quinoline-3-carboxylic acid
Molecular Weight (g/mol) 173.17
Formula Weight 173.0477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.