missing translation for 'onlineSavingsMsg'
Learn More

3-Quinuclidinone hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 15234634 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15234634 500 g Plastic Bottle 500g
10306830 25 g Plastic Bottle 25g
10285420 100 g Plastic Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15234634 Supplier Thermo Scientific Acros Supplier No. 132265000
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-65-3
Molecular Formula C7H11NO·HCl
Molecular Weight (g/mol) 161.63
MDL Number MFCD00137391
InChI Key RFDPHKHXPMDJJD-UHFFFAOYSA-N
Synonym 3-quinuclidinone hydrochloride, quinuclidin-3-one hydrochloride, 3-quinuclidone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one hydrochloride, 3-quinuclidnone hydrochloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride, 1-aza-3-oxobicyclo 2.2.2 octane, quinuclidin-3-one, chloride, 1-azabicyclo 2.2.2 octan-3-one, hydrochloride 1:1
PubChem CID 102019
IUPAC Name 1-azabicyclo[2.2.2]octan-3-one;hydrochloride
SMILES C1CN2CCC1C(=O)C2.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White
Loss on Drying 0.5% max. (100°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (On dry substance) (Argentometry)
Packaging Plastic Bottle
Quantity 500 g
Solubility Information Solubility in water: soluble
Formula Weight 161.63
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Quinuclidinone hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Quinuclidinone hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.