missing translation for 'onlineSavingsMsg'
Learn More

3(R,S)-Hydroxy Desogestrel, TRC

Product Code. 30397470
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397470 100 mg -
30389308 50 mg 50mg
30402244 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397470 Supplier Toronto Research Chemicals Supplier No. TRCH939000100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869627-85-0
Molecular Formula C22H30O2
Molecular Weight (g/mol) 326.47
Synonym 3-Hydroxy-desogestrel, 18,19-Dinorpregn-4-en-20-yne-3,17-diol, 13-ethyl-11-methylene-, (17α)- (9CI), 3(R,S)-Hydroxy desogestrel, 3-Hydroxydesogestrel, Desogestrel Related Compound B (USP)
SMILES CC[C@]12CC(=C)[C@H]3[C@@H](CCC4=CC(O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H30O2/c1-4-21-13-14(3)20-17-9-7-16(23)12-15(17)6-8-18(20)19(21)10-11-22(21,24)5-2/h2,12,16-20,23-24H,3-4,6-11,13H2,1H3/t16?,17-,18-,19?,20?,21-,22?/m0/s1
Formula Weight 326.2246
Chemical Name or Material 3(R,S)-Hydroxy Desogestrel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.