missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Rhodanineacetic Acid, TRC
Specifications
Specifications
| CAS | 5718-83-2 |
| Molecular Formula | C5H5NO3S2 |
| Quantity | 5 g |
| Synonym | 4-Oxo-2-Thioxo-3-thiazolidineacetic Acid, (4-Oxo-2-thioxo-1,3-thiazolidin-3-yl)acetic Acid, (4-Oxo-2-thioxothiazolidin-3-yl)acetic Acid, 2-(4-Oxo-2-thioxothiazolidin-3-yl)acetic Acid, 3-(Carboxymethyl)rhodanine, 4-Oxo-2-thioxothiazolidine-3-acetic Acid, N-(Carboxymethyl)rhodanine, NSC 40450, Rhodanine-3-ethanoic Acid, Rhodanine-N-acetic Acid |
| InChI Formula | InChI=1S/C5H5NO3S2/c7-3-2-11-5(10)6(3)1-4(8)9/h1-2H2,(H,8,9) |
| SMILES | OC(=O)CN1C(=O)CSC1=S |
| IUPAC Name | 2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid |
| Molecular Weight (g/mol) | 191.23 |
| Formula Weight | 190.9711 |
| Chemical Name or Material | 3-Rhodanineacetic Acid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?