missing translation for 'onlineSavingsMsg'
Learn More

3-(tert-Butyl)-1-methyl-1H-pyrazol-5-amine, 97%, Thermo Scientific™

Product Code. 10702113
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10702113 1 g Amber Glass Bottle 1g
10468192 10 g Amber Glass Bottle 10g
10377544 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10702113 Supplier Thermo Scientific Maybridge Supplier No. KM00494DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 118430-73-2
Molecular Formula C8H15N3
Molecular Weight (g/mol) 153.23
MDL Number MFCD00068002
InChI Key XSCDSAMVQLKDNI-UHFFFAOYSA-N
Synonym 5-amino-3-tert-butyl-1-methylpyrazole, 3-tert-butyl-1-methyl-1h-pyrazol-5-amine, 5-tert-butyl-2-methyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 3-1,1-dimethylethyl-1-methyl, 5-amino-3-tert-butyl-1-methyl-1h-pyrazole, 3-tert-butyl-1-methylpyrazole-5-ylamine, acmc-1cins, maybridge4_002215, 5-amino-1-methyl-3-tert-butylpyrazole
PubChem CID 2735287
SMILES CN1N=C(C=C1N)C(C)(C)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 5-tert-butyl-2-methylpyrazol-3-amine
Formula Weight 153.23
Percent Purity 97%
Chemical Name or Material 3-(tert-Butyl)-1-methyl-1H-pyrazol-5-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.