missing translation for 'onlineSavingsMsg'
Learn More

3-[(Tetrahydrofuran-2-ylmethyl)amino]propanenitrile, 97%, Thermo Scientific™

Product Code. 10304532
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10304532 1 g Amber Glass Bottle 1g
10651103 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10304532 Supplier Thermo Scientific Maybridge Supplier No. KM09269DA
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 90322-18-2
Molecular Formula C8H14N2O
Molecular Weight (g/mol) 154.21
MDL Number MFCD00121811
InChI Key ZSKZHSOMHSHFDF-UHFFFAOYNA-N
Synonym 3-tetrahydrofuran-2-ylmethyl amino propanenitrile, 3-oxolan-2-ylmethyl amino propanenitrile, 3-tetrahydro-2-furanylmethyl amino-propanenitrile, 3-tetrahydrofuran-2-yl methyl amino propanenitrile, 3-oxolan-2-ylmethylamino propanenitrile, 3-oxolan-2-yl methyl amino propanenitrile, 3-tetrahydro-furan-2-ylmethyl-amino-propionitrile, 3-tetrahydrofuran-2-yl ;methyl ;amino ;propanenitrile, propanenitrile,3-tetrahydro-2-furanyl methyl amino
PubChem CID 2779328
IUPAC Name 3-(oxolan-2-ylmethylamino)propanenitrile
SMILES N#CCCNCC1CCCO1

Specifications

CAS Min % 97.0
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 3-[(Tetrahydrofuran-2-ylmethyl)amino]propanenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.