missing translation for 'onlineSavingsMsg'
Learn More

3-Thien-3-ylaniline, ≥97%, Thermo Scientific™

Product Code. 10612875
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10612875 5 g 5g
10563135 250 mg 250mg
10225324 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10612875 Supplier Thermo Scientific Maybridge Supplier No. CC48414DE
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 161886-96-0
Molecular Formula C10H9NS
Molecular Weight (g/mol) 175.25
MDL Number MFCD06740169
InChI Key SHUODWYRFVFGOA-UHFFFAOYSA-N
Synonym 3-thien-3-ylaniline, 3-thiophen-3-yl aniline, 3-3-aminophenyl thiophene, 3-thiophen-3-yl-phenylamine, 3-3-thienyl aniline, 3-3-thienyl phenylamine, 3-thiophen-3-ylphenylamine, benzenamine,3-3-thienyl, 3-thiophen-3-yl benzenamine, benzenamine, 3-3-thienyl
PubChem CID 4130337
SMILES NC1=CC=CC(=C1)C1=CSC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
IUPAC Name 3-thiophen-3-ylaniline
Formula Weight 175.25
Percent Purity 97%
Chemical Name or Material 3-Thien-3-ylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.