missing translation for 'onlineSavingsMsg'
Learn More

3-Thiophen-2-yl-piperazine-1-carboxylic acid ethyl ester, Thermo Scientific™

Product Code. 10507780
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10507780 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10507780 Supplier Thermo Scientific Maybridge Supplier No. MAY00167DA
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 85803-50-5
Packaging Amber Glass Bottle
Molecular Formula C11H16N2O2S
Quantity 1 g
Synonym ethyl 3-thiophen-2-yl piperazine-1-carboxylate, ethyl 3-2-thienyl piperazine-1-carboxylate, 3-thiophen-2-yl-piperazine-1-carboxylic acid ethyl ester, acmc-20amyi, ethyl 3-2-thienyl piperazinecarboxylate, ethyl 3-2-thienyl piperazine-carboxylate, 3-thien-2-yl-piperazine-1-carboxylic acid ethyl ester, 3-thiophen-2-yl-piperazine-1-carboxylic acidethyl ester, 3-thiophen-2-ylpiperazine-1-carboxylic acid ethyl ester, ethyl 3-thiophen-2-yl-piperazine-1-carboxylate
InChI Key YNIWHEQLSSKQCX-UHFFFAOYSA-N
SMILES CCOC(=O)N1CCNC(C1)C2=CC=CS2
IUPAC Name ethyl 3-thiophen-2-ylpiperazine-1-carboxylate
Molecular Weight (g/mol) 240.321
PubChem CID 2771741
Formula Weight 240.32
Percent Purity 95%
Chemical Name or Material 3-thiophen-2-yl-piperazine-1-carboxylic acid ethyl ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.