missing translation for 'onlineSavingsMsg'
Learn More

3-Thiopheneethanol, 98%, Thermo Scientific Chemicals

Product Code. 11358166
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358166 5 g 5g
11348166 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358166 Supplier Thermo Scientific Alfa Aesar Supplier No. L15138.06
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

3-Thiopheneethanol is used in the synthesis of various ether and ester derivatives such as 3-(2-(benzyloxy)ethyl)thiophene, 3-[2-((triphenylmethyl)oxy)ethyl]thiophene, 3-[2-((trimethylsilyl)oxy)ethyl]thiophene, 3-[2-((dimethyl-tert-butylsilyl)oxy)ethyl]thiophene, 3-(2-acetoxyethyl)thiophene and 3-(2-(benzoyloxy)ethyl)thiophene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiopheneethanol is used in the synthesis of various ether and ester derivatives such as 3-(2-(benzyloxy)ethyl)thiophene, 3-[2-((triphenylmethyl)oxy)ethyl]thiophene, 3-[2-((trimethylsilyl)oxy)ethyl]thiophene, 3-[2-((dimethyl-tert-butylsilyl)oxy)ethyl]thiophene, 3-(2-acetoxyethyl)thiophene and 3-(2-(benzoyloxy)ethyl)thiophene

Solubility
Sparingly soluble in water(0.059 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13781-67-4
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00009766
InChI Key YYPNNBPPDFTQFX-UHFFFAOYSA-N
Synonym 2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415
PubChem CID 83731
IUPAC Name 2-thiophen-3-ylethanol
SMILES OCCC1=CSC=C1

Specifications

Density 1.159
Boiling Point 110°C to 111°C (14 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.553
Quantity 5 g
Beilstein 107080
Solubility Information Sparingly soluble in water(0.059g/L at 25°C).
Formula Weight 128.19
Percent Purity 98%
Chemical Name or Material 3-Thiopheneethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.