missing translation for 'onlineSavingsMsg'
Learn More

3-Tolylboronic acid, 97%

Product Code. 10778362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10778362 1 g Glass bottle 1g
10778552 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10778362 Supplier Thermo Scientific Acros Supplier No. 344420010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17933-03-8 | C7H9BO2 | 135.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17933-03-8
Molecular Formula C7H9BO2
Molecular Weight (g/mol) 135.96
MDL Number MFCD00040198
InChI Key BJQCPCFFYBKRLM-UHFFFAOYSA-N
Synonym 3-tolylboronic acid, m-tolylboronic acid, 3-methylphenyl boronic acid, 3-methylbenzeneboronic acid, 3-methyl phenyl boronic acid, 3-tolyboronic acid, 3-methylphenyl boranediol, m-methylphenylboronic acid, m-tolyl boronic acid
PubChem CID 2733950
IUPAC Name (3-methylphenyl)boronic acid
SMILES CC1=CC=CC(=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 160.0°C to 162.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3C6H4B(OH)2
Quantity 1 g
Solubility Information Solubility in water: +/- 2.5% in water.
Formula Weight 135.96
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material 3-Tolylboronic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Tolylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.